missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, NF, 85-88%, Spectrum™ Chemical

PH175, 7664-38-2, H3PO4

Supplier:  Spectrum Chemical Mfg Cor PH1751LT

 View more versions of this product

Catalog No. 18605180

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Phosphoric Acid, NF - All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7664-38-2
Amber Glass Bottle
H3O4P
OP(O)(O)=O
97.99
NF
1
1 L
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
85 to 88%

Chemical Identifiers

7664-38-2
97.99
phosphoric acid
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
OP(O)(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions