missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, FCC, 85%, Spectrum™ Chemical

P1097, 7664-38-2, H3PO4

$601.00 - $14558.17

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605173
View Documents
Spectrum Chemical Mfg Cor
P1097500MLGL
500 mL Amber Glass Bottle
Each for $601.00
Only null left
 
18605168
View Documents
Spectrum Chemical Mfg Cor
P10971LT
1 L Amber Glass Bottle
Each for $726.13
Only null left
 
18605169
View Documents
Spectrum Chemical Mfg Cor
P109725LTPV
2.5 L Poly-Coated Glass Bottle
Each for $1,225.33
Only null left
 
18605174
View Documents
Spectrum Chemical Mfg Cor
P1097500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $2,682.93
Only null left
 
18605170
View Documents
Spectrum Chemical Mfg Cor
P10974LT
4 L Poly-Coated Glass Bottle
Each for $1,422.77
Only null left
 
18605171
View Documents
Spectrum Chemical Mfg Cor
P10971LTCS6
6 x 1 L Amber Glass Bottle
Case of 6 Each for $3,130.17
Only null left
 
18605175
View Documents
Spectrum Chemical Mfg Cor
P109720LTBL
20 L Poly Drum
Each for $4,989.40
Only null left
 
18605176
View Documents
Spectrum Chemical Mfg Cor
P1097200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Phosphoric Acid, 85 Percent, FCC is used to acidify foods and beverages such as various colas and jams. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7664-38-2
97.99
phosphoric acid
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
OP(O)(O)=O

Specifications

7664-38-2,7732-18-5
Amber Glass Bottle
H3O4P
OP(O)(O)=O
97.99
FCC
85 to 88%,12 to 15%
500 mL
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
85%
Phosphoric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'