Learn More
Phosphoric Acid, Certified, 10.0% (v/v) ±0.2% (v/v), LabChem™
Supplier: LabChem LC186502
Specifications
| Phosphoric Acid | |
| 7664-38-2,7732-18-5 | |
| Passes Test | |
| Traceable to NIST | |
| 1 L | |
| Liquid | |
| UN1805 | |
| Soluble in water | |
| OP(=O)(O)O | |
| 97.994 | |
| CHEBI:26078 | |
| 10.0% ±0.2% (v/v) |
| Colorless | |
| 90,10 | |
| Poly Bottle | |
| Phosphorus oxides; Thermal decomposition generates: Corrosive vapors | |
| H3O4P | |
| H3PO4 | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 1004 | |
| 97.1 | |
| Certified |
Chemical Identifiers
| 7664-38-2 | |
| 97.994 | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| CHEBI:26078 | |
| OP(=O)(O)O |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| 1004 | |
| phosphoric acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature