missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, ACS Grade, ≥ 85.0% (w/w), LabChem™

Catalog No. LC186401
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 2.5 L
Packaging:
Amber Glass
HDPE Bottle
Poly Bottle
PVC Coated
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC186401 500 mL Amber Glass
LC186403 2.5 L HDPE Bottle
LC186403S 2.5 L Poly Bottle
LC1864036S 6 x 2.5 L PVC Coated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC186401 Supplier LabChem Supplier No. LC186401

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
Melting Point 21°C
Boiling Point 158°C
CAS Max % 100
Density 1,685kg/m3
Identification Passes Test
Packaging Amber Glass
Reducing Substances Passes Test
Specific Gravity 1685kg/m3
Decomposition Information Phosphorus oxides; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Linear Formula H3PO4
UN Number UN1805
Solubility Information Soluble in water
Formula Weight 97.1
Percent Purity ≥85.0% (w/w)
Grade ACS
Chemical Name or Material Phosphoric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-633-2
Recommended Storage Room Temperature