missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, ACS, 85%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P1095200LTBL
Specifications
| 7664-38-2,7732-18-5 | |
| 1e-05 | |
| To Pass Test | |
| H3O4P | |
| OP(O)(O)=O | |
| 97.99 | |
| ACS |
| 1 | |
| Drum | |
| 200 L | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 85% |
Chemical Identifiers
| 7664-38-2 | |
| 97.99 | |
| phosphoric acid |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| OP(O)(O)=O |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.