Learn More
Phosphoric acid, 85% w/w aq. soln., ACS
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Supplier: Thermo Scientific Chemicals 033266A1
Description
Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and in leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion into a water soluble phosphate compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Phosphoric acid | |
| 7664-38-2 | |
| Viscous Liquid | |
| 85% w/w aq. soln. | |
| MFCD00011340 | |
| Miscible with water,ethanol and mixture of ether: alcohol (3:1). | |
| OP(=O)(O)O | |
| 97.994 | |
| CHEBI:26078 | |
| ACS | |
| Colorless viscous liquid | |
| 1.7 |
| 19°C to 21°C | |
| Colorless | |
| 1 kg | |
| H3O4P | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 1004 | |
| 98 | |
| 1.431 | |
| 23 hPa (17mm Hg) at 20°C |
Chemical Identifiers
| 7664-38-2 | |
| 97.994 | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| 1004 | |
| phosphoric acid |
| H3O4P | |
| MFCD00011340 | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| CHEBI:26078 | |
| OP(=O)(O)O |
Safety and Handling
P234-P264b-P280-P301+P330+P331-P303+P361+P353-P305+P351+P338-P310-P342+P311-P363-P390-P501c
H290-H314
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-633-2
RTECSNumber : TB6300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only