missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric acid, 85% w/w aq. soln., ACS

Catalog No. AA3326622
Change view
Click to view available options
Quantity:
100 g
1 kg
4 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3326622 100 g
AA33266A1 1 kg
AA33266D6 4 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3326622 Supplier Thermo Scientific Chemicals Supplier No. 03326622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7664-38-2 | H3O4P | 97.994 g/mol

Phosphoric acid is used mainly in fertilizers. It is used as an external standard for phosphorus-31 Nuclear magnetic resonance (NMR); as an electrolyte in phosphoric acid fuel cells and as a catalyst in the hydration of alkenes to produce alcohols, predominantly ethanol. It finds application as a buffer agent in biology and chemistry; as a solution for anodizing and as a cleaner by construction trades to remove mineral deposits, cementitious smears and hard water stains. It is also used as a pH adjuster in cosmetics and skin-care products and a dispersing agent in detergents and in leather treatment. It plays an important role in the preparation of albumin derivatives. It is an important starting material used in the preparation of disodium hydrogen phosphate, trisodium phosphate, calcium dihydrogen phosphate and monohydrogen phosphate. It serves as an acidic fruitlike flavoring in food products. It is widely used to remove rust in steel tools or surfaces by converting the rusted ion into a water soluble phosphate compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Melting Point 19°C to 21°C
Color Colorless
Physical Form Viscous Liquid
Quantity 100 g
Assay Percent Range 85% w/w aq. soln.
Solubility Information Miscible with water,ethanol and mixture of ether: alcohol (3:1).
Formula Weight 98
Grade ACS
Refractive Index 1.431
Appearance Colorless viscous liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.70g/cm3 at 20°C
Chemical Name or Material Phosphoric acid
Show More Show Less
Hazard Category H290-H314
Precautionary Statement P234-P264b-P280-P301+P330+P331-P303+P361+P353-P305+P351+P338-P310-P342+P311-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-633-2
RTECSNumber TB6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only