missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid 85% LM Prime, Transene

Catalog No. 502744713
Encompass
Catalog No. 502744713 Supplier Doe and Ingalls Management LLC Supplier No. T0701615GAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable For Cleaning And Etching In Electronic Applications

Phosphoric acid, also known as orthophosphoric acid, is a weak triprotic acid. At 85%, it is a colorless and odorless viscous liquid. Semiconductor grade indicates that the solvent is of sufficient purity for the semiconductor manufacturing and electronic industries, having only trace levels of impurities.

  • Common Uses and Applications:
  • Etching of semiconductors
  • Sanitizing agent for silicon wafers
  • Rust converter
  • Anti-rust agent
  • Cleaning agent
  • Industries
  • Electronic industry
  • Safety and Shipping Information DOT: Phosphoric Acid, 8, UN1805, PG III

Chemical Identifiers

CAS 7664-38-2
Molecular Weight (g/mol) 98 g/mol

Specifications

Packaging HDPE
Quantity 1 gal.
Percent Purity 85%
Grade Electronic
Chemical Name or Material Phosphoric Acid