missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 0.085% in Acetonitrile, Reagents

Catalog No. 01334142
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334142 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334142 Supplier Labchem Inc Supplier No. CP2230004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphoric Acid, 0.085% in Acetonitrile Manufactured with HPLC components, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-05-8 , 7664-38-2
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Boiling Point 82°C
Packaging Glass Bottle
Specific Gravity 0.7875
Quantity 4 L
Physical Form Liquid
Percent Purity 0.08%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 0.085% in Acetonitrile Manufactured with HPLC components