Learn More
Phosphonoacetic acid, 98+%
CAS: 4408-78-0 | C2H5O5P | 140.03 g/mol
Supplier: Thermo Scientific Chemicals A1211714
Description
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Phosphonoacetic acid | |
| 139°C to 146°C | |
| C2H5O5P | |
| 25 g | |
| 1764355 | |
| phosphonoacetic acid, fosfonet, phosphonoacetate, acetic acid, phosphono, fosfonet sodium, phosphonacetic acid, carboxymethanephosphonic acid, fosfonoacetic acid, disodium phosphonoacetate, lopac-p-6909 | |
| XUYJLQHKOGNDPB-UHFFFAOYSA-N | |
| 2-phosphonoacetic acid | |
| 546 | |
| 140.04 |
| 4408-78-0 | |
| Odorless | |
| MFCD00004311 | |
| UN3261 | |
| Hygroscopic | |
| Soluble in water. | |
| OC(=O)CP(O)(O)=O | |
| 140.03 | |
| CHEBI:15732 | |
| ≥98% |
Chemical Identifiers
| 4408-78-0 | |
| 140.03 | |
| XUYJLQHKOGNDPB-UHFFFAOYSA-N | |
| 546 | |
| 2-phosphonoacetic acid |
| C2H5O5P | |
| MFCD00004311 | |
| phosphonoacetic acid, fosfonet, phosphonoacetate, acetic acid, phosphono, fosfonet sodium, phosphonacetic acid, carboxymethanephosphonic acid, fosfonoacetic acid, disodium phosphonoacetate, lopac-p-6909 | |
| CHEBI:15732 | |
| OC(=O)CP(O)(O)=O |
Safety and Handling
Causes severe skin burns and eye damage
Skin corrosion/irritation (category 1)
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 224-558-1
RTECSNumber : AJ3278000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only