missing translation for 'onlineSavingsMsg'
Learn More

Phosphonitrilic Chloride Trimer 98.0+%, TCI America™

Catalog No. p-7124512
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058425G 25 g
C0584250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C058425G Supplier TCI America Supplier No. C058425G
Only null left

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.642
MDL Number MFCD00006474
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Melting Point 113°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 1759
Formula Weight 347.64
Percent Purity ≥98.0% (GC)
Chemical Name or Material Phosphonitrilic Chloride Trimer
EINECSNumber (1)-0652
TSCA Yes