Learn More
Phosphonitrilic chloride trimer, 98%
CAS: 940-71-6 | Cl6N3P3 | 347.642 g/mol
Supplier: Thermo Scientific Chemicals A1898622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 127°C (13mmHg) | |
| 111°C to 115°C | |
| 100 g | |
| MFCD00006474 | |
| phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
| N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl | |
| 347.642 | |
| 347.66 | |
| Moisture sensitive |
| Phosphonitrilic chloride trimer | |
| 940-71-6 | |
| Cl6N3P3 | |
| UN3260 | |
| UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
| 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
| 220225 | |
| 98% | |
| 1.98 |
Chemical Identifiers
| 940-71-6 | |
| 347.642 | |
| UBIJTWDKTYCPMQ-UHFFFAOYSA-N | |
| 220225 | |
| N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl |
| Cl6N3P3 | |
| MFCD00006474 | |
| phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer | |
| 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 213-376-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only