missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic Acid Hydrate, TCI America™
Supplier: TCI America P1910100G
Specifications
| Phosphomolybdic Acid Hydrate | |
| 90°C | |
| 100 g | |
| Crystal-Powder at 20°C | |
| 3085 | |
| FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
| 12-Molybdophosphoric acid | |
| 1825.48 |
| Yellow | |
| 51429-74-4 | |
| H3Mo12O40P | |
| MFCD00149913 | |
| Molybdo(VI)phosphoric Acid | |
| [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O | |
| 1825.40 |
Chemical Identifiers
| 51429-74-4 | |
| 1825.40 | |
| FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
| 12-Molybdophosphoric acid |
| H3Mo12O40P | |
| MFCD00149913 | |
| Molybdo(VI)phosphoric Acid | |
| [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O |
Safety and Handling
EINECSNumber : (1)-0714
TSCA : Yes