missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic Acid hydrate, ACS Reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Color | Yellow |
| Heavy Metals (as Pb) | 50ppm max. |
| Packaging | Glass Bottle |
| Linear Formula | H3[P(Mo3O10)4]·xH2O |
| Quantity | 500 g |
| Physical Form | Crystals |
| Merck Index | 14, 7342 |
| Synonym | Molybdophosphoric Acid |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and ether |
| Formula Weight | 1825.25 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. May intensify fire; oxidizer. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only