missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic acid, ammonium salt hydrate

Catalog No. AC317100500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317100500 50 g Plastic bottle
AC317102500 250 g Plastic bottle
AC317100010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC317100500 Supplier Thermo Scientific Chemicals Supplier No. 317100500

CAS: 54723-94-3 | H80Mo12N3O41P-6 | 1960.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 54723-94-3
Molecular Formula H80Mo12N3O41P-6
Molecular Weight (g/mol) 1960.994
MDL Number MFCD00150423
InChI Key BIQVMZMYTFVCJZ-UHFFFAOYSA-H
Synonym ammonium phosphomolybdate hydrate
PubChem CID 131664298
IUPAC Name azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
SMILES N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]

Specifications

Color Yellow
Packaging Plastic bottle
Quantity 50 g
Physical Form Powder
Linear Formula (NH4)3PMo12O40·xH2O
Solubility Information Solubility in water: insoluble.
Formula Weight 1876.35
Chemical Name or Material Phosphomolybdic acid, ammonium salt hydrate
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only