missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphocholine Chloride Calcium Salt Tetrahydrate 98.0+%, TCI America™
Supplier: TCI America P0274250G
Specifications
| Phosphocholine Chloride Calcium Salt Tetrahydrate | |
| White | |
| MFCD00149983 | |
| Phosphorylcholine Chloride Calcium Salt | |
| C[N+](C)(C)CCOP(=O)(O)O.O.O.O.O.[Cl-].[Ca] | |
| 331.74 | |
| 257.66 | |
| Crystalline Powder |
| 72556-74-2 | |
| C5H23CaClNO8P | |
| 250 g | |
| YFRZEXLBVHTMGV-UHFFFAOYSA-N | |
| calcium;trimethyl(2-phosphonooxyethyl)azanium;chloride;tetrahydrate | |
| 131858236 | |
| ≥98.0% (T) |
Chemical Identifiers
| 72556-74-2 | |
| 331.74 | |
| YFRZEXLBVHTMGV-UHFFFAOYSA-N | |
| 131858236 | |
| C[N+](C)(C)CCOP(=O)(O)O.O.O.O.O.[Cl-].[Ca] |
| C5H23CaClNO8P | |
| MFCD00149983 | |
| Phosphorylcholine Chloride Calcium Salt | |
| calcium;trimethyl(2-phosphonooxyethyl)azanium;chloride;tetrahydrate |
Safety and Handling
EINECSNumber : (2)-1954&(9)-0261
TSCA : No