missing translation for 'onlineSavingsMsg'
Learn More

Phosphocholine Chloride Calcium Salt Tetrahydrate 98.0+%, TCI America™

Catalog No. P027425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P027425G 25 g
P0274250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P027425G Supplier TCI America Supplier No. P027425G

Chemical Identifiers

CAS 72556-74-2
Molecular Formula C5H23CaClNO8P
Molecular Weight (g/mol) 331.74
MDL Number MFCD00149983
InChI Key YFRZEXLBVHTMGV-UHFFFAOYSA-N
Synonym Phosphorylcholine Chloride Calcium Salt
PubChem CID 131858236
IUPAC Name calcium;trimethyl(2-phosphonooxyethyl)azanium;chloride;tetrahydrate
SMILES C[N+](C)(C)CCOP(=O)(O)O.O.O.O.O.[Cl-].[Ca]

Specifications

Color White
Quantity 25 g
Formula Weight 257.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phosphocholine Chloride Calcium Salt Tetrahydrate
EINECSNumber (2)-1954&(9)-0261
TSCA No