missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphatidylethanol, ≥98%, MP Biomedicals™
A pathological phospholipid, formed via the action of phospholipase D only in the presence of ethanol
$657.88 - $657.88
Chemical Identifiers
| Molecular Formula | C39H75O8P |
|---|---|
| Synonym | 1-Palmitoyl-2-oleoyl-sn-3-phosphoethanol, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15898310
|
MP Biomedicals Inc
0215898310 |
10 mg |
Each for $657.88
|
|
|||||
Chemical Identifiers
| C39H75O8P | |
| 1-Palmitoyl-2-oleoyl-sn-3-phosphoethanol, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol |
Specifications
| ≥98% | |
| 10 mg | |
| 703 | |
| Phosphatidylethanol |
| C39H75O8P | |
| 1-Palmitoyl-2-oleoyl-sn-3-phosphoethanol, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol | |
| Viscous Liquid or Waxy Solid |