missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Phosphate-buffered saline (PBS, 10X), pH 7.4, for Western blot
For Western blot washing
$395.19
Specifications
| Color | Colorless |
|---|---|
| For Use With (Application) | For Western blot washing. |
| pH | 7.4 |
| Product Type | Phosphate-buffered Saline |
| Catalog Number | Mfr. No. | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimQuantity | Price | Quantity & Availability | |||||
|
AAJ60801K3
|
Thermo Scientific™
J60801K3 |
N/A
|
N/A | ||||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAJ60801K7
|
Thermo Scientific™
J60801K7 |
Each for $395.19
|
|
||||||
|
AAJ60801K2
|
Thermo Scientific™
J60801K2 |
N/A
|
N/A | ||||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Colorless | |
| For Western blot washing. | |
| 7.4 | |
| Phosphate-buffered Saline |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only