missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution, MilliporeSigma™ Supelco™

Catalog No. 111005316
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111005316 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005316 Supplier Merck Emd Millipore Supplier No. 76847100MLF
Only null left

Specifications

Density 1.0 g/mL (at 20°C)
pH 7.2 ±0.2
Quantity 100 mL
Grade BioUltra
Concentration or Composition (by Analyte or Components) >0.022 M NaH2 PO4; >0.044 M Na2 HPO4