missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer Solution For Chlorine Determination (DPD Methods), Reagents

Catalog No. 01334147
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334147 4 L
01334146 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334147 Supplier Labchem Inc Supplier No. CP2109004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Buffer Solution For Chlorine Determination (DPD Methods), Reagent, use for Water and wastewater analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 4 L
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible