missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, pH 8.8, Ricca Chemical

Catalog No. R5814500150
Change view
Click to view available options
Quantity:
150 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5814500150 150 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5814500150 Supplier Ricca Chemical Company Supplier No. R5814500150A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Phosphate Buffer
Name Note pH 8.8
CAS 7758-11-4, 1310-58-3
CAS Min % 96.0, 1.67, 0.02
CAS Max % 100.0, 1.74, 0.02
Quantity 150 mL
Molecular Formula HK2O4P
Packaging Natural Poly Bottle
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Molecular Weight (g/mol) 174.17