missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, pH 7, Reagents

Catalog No. 01334144
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334144 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334144 Supplier Labchem Inc Supplier No. CP2115001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Buffer, pH 7, ASTM D 1094, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 7
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Sterility Non-sterile
Solubility Information Completely miscible