missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer pH 6.2 to 6.5, For Chlorine, Chlorine Dioxide, Certified, LabChem™

Catalog No. LC185402
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC185402 1 L
LC185401 500 mL
LC185404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC185402 Supplier LabChem Supplier No. LC185402
Only null left

Specifications

Color Clear, colorless
pH Range 6.2 to 6.5
Format Liquid
Packaging Poly Bottle
Quantity 1 L
Storage Requirements Room Temperature
Type Phosphate Buffer pH 6.2 to 6.5