missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffer, pH 3.15, Reagents

Catalog No. 01334157
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334157 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334157 Supplier Labchem Inc Supplier No. CP20940020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Buffer, pH 3.15, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
pH 3.15
Physical Form Liquid
Quantity 20 L
Packaging Cubitainer
Sterility Non-sterile
Solubility Information Completely miscible