missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 9.0

Catalog No. AAJ62345AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62345AP 500 mL
AAJ62345AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62345AP Supplier Thermo Scientific™ Supplier No. J62345AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 7558-80-7
Concentration 0.5M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle
Molecular Weight (g/mol) 119.976