missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 8.5

Catalog No. AAJ61086AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61086AK 250 mL
AAJ61086AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61086AK Supplier Thermo Scientific™ Supplier No. J61086AK

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 7558-80-7
Concentration 0.5M buffer soln.
Physical Form Liquid
Quantity 250 mL
Packaging Plastic Bottle
Molecular Weight (g/mol) 119.976