Learn More
Enzo Life Sciences alphaB Crystallin (pSer59) polyclonal antibody (50 g)
Supplier: Enzo ADISPA227D
Host: Rabbit. Immunogen: Synthetic peptide corresponding to to aa 54-64 of of human B-crystallin phosphorylated at Ser59. UniProt: P02511. Source: Purified from rabbit serum. Species Reactivity: Mouse, Rat, Sheep, Porcine, Bovine. Applications: IP, WB. Recommended dilutions: Western Blot (ECL, 1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In sodium phosphate buffer, pH 7.0, containing 0.1% sodium azide, 3% BSA, and 50% glycerol.