missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Phorbol-12,13-dibutyrate, Calbiochem™,

Catalog No. 5243901MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Alu drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5243901MG 1 mg Alu drum
5243905MG 5 mg Alu drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5243901MG Supplier MilliporeSigma™ Supplier No. 5243901MG

Chemical Identifiers

CAS 37558-16-0
Molecular Formula C28H40O8
Molecular Weight (g/mol) 504.62
InChI Key BQJRUJTZSGYBEZ-YVQNUNKESA-N
Synonym phorbol 12,13-dibutyrate, pdbu, phorbol dibutyrate, phorbol 12,13-dibutanoate, phorbol-12,13-dibutyrate, unii-67mx82cl58, chembl27768, 1ar,1bs,4ar,7as,7bs,8r,9r,9as-4a,7b-dihydroxy-3-hydroxymethyl-1,1,6,8-tetramethyl-5-oxo-1,1a,1b,4,4a,5,7a,7b,8,9-decahydro-9ah-cyclopropa 3,4 benzo 1,2-e azulene-9,9a-diyl dibutanoate, butanoic acid, 1ar,1bs,4ar,7as,7bs,8r,9r,9as-1,1a,1b,4,4a,5,7a,7b,8,9-decahydro-4a,7b-dihydroxy-3-hydroxymethyl-1,1,6,8-tetramethyl-5-oxo-9ah-cyclopropa 3,4 benz 1,2-e azulene-9,9a-diyl ester, butanoic acid, 1,1a,1b,4,4a,5,7a,7b,8,9-decahydro-4a,7b-dihydroxy-3-hydroxymethyl-1,1,6,8-tetramethyl-5-oxo-9ah-cyclopropa 3,4 benz 1,2-e azulene-9,9a-diyl ester, 1ar-1a-alpha,1b-beta,4a-beta,7a-alpha,7b-alpha,8-alpha,9-beta,9a-alpha
PubChem CID 37783
ChEBI CHEBI:17598
SMILES CCCC(=O)OC1C(C2(C(C=C(CC3(C2C=C(C3=O)C)O)CO)C4C1(C4(C)C)OC(=O)CCC)O)C

Specifications

Color White
Packaging Alu drum
Quantity 1 mg
Percent Purity ≥99% by HPLC
Physical Form Crystalline Solid