missing translation for 'onlineSavingsMsg'
Learn More

Phlorizin Hydrate 97.0+%, TCI America™

$254.11 - $698.84

Chemical Identifiers

CAS 60-81-1
Molecular Formula C21H24O10
Molecular Weight (g/mol) 436.413
MDL Number MFCD00006591
InChI Key IOUVKUPGCMBWBT-QNDFHXLGSA-N
Synonym phlorizin, phloridzin, phlorhizin, phlorizoside, floridzin, phlorrhizin, phloretin 2'-glucoside, phloridzosid, phlorrhizen, phlorizine
PubChem CID 6072
ChEBI CHEBI:8113
IUPAC Name 1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
SMILES C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P02481G
View Documents
TCI America
P02481G
1 g
Each for $254.11
Only null left
 
P02485G
View Documents
TCI America
P02485G
5 g
Each for $698.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60-81-1
436.413
IOUVKUPGCMBWBT-QNDFHXLGSA-N
6072
1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
C21H24O10
MFCD00006591
phlorizin, phloridzin, phlorhizin, phlorizoside, floridzin, phlorrhizin, phloretin 2'-glucoside, phloridzosid, phlorrhizen, phlorizine
CHEBI:8113
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Specifications

60-81-1
MFCD00006591
phlorizin, phloridzin, phlorhizin, phlorizoside, floridzin, phlorrhizin, phloretin 2'-glucoside, phloridzosid, phlorrhizen, phlorizine
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O
436.413
CHEBI:8113
≥97.0% (T)
Phlorizin Hydrate
C21H24O10
1 g
IOUVKUPGCMBWBT-QNDFHXLGSA-N
1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
6072
436.41
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UC2080000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'