missing translation for 'onlineSavingsMsg'
Learn More

Phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. aah58659ad
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58659AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58659AD Supplier Thermo Scientific Chemicals Supplier No. H58659AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38111-44-3 | C6H5BrZn | 222.39 g/mol

It is used in the cross-coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the cross-coupling reaction.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate

Chemical Identifiers

CAS 38111-44-3
Molecular Formula C6H5BrZn
Molecular Weight (g/mol) 222.39
MDL Number MFCD01319861
InChI Key HMFWJMIDCMEJHO-UHFFFAOYSA-M
Synonym phenylzinc bromide, zinc, bromophenyl, phenylzinc bromide solution, benzene,bromozinc 1+, phenylzinc bromide 0.5 m in tetrahydrofuran, phenylzinc bromide solution, 0.5 m in thf, phenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 16213817
IUPAC Name zinc;benzene;bromide
SMILES [Zn++].[Br-].C1=CC=[C-]C=C1

Specifications

Density 0.99
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 222.39
Chemical Name or Material Phenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only