missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylsilane, 98%

Catalog No. AAL0287614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0287614 25 g
AAL0287606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0287614 Supplier Thermo Scientific Chemicals Supplier No. L0287614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 768-32-1 | C9H14Si | 150.30 g/mol

It is employed as intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical and organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 768-32-1
Molecular Formula C9H14Si
Molecular Weight (g/mol) 150.30
MDL Number MFCD00008271
InChI Key KXFSUVJPEQYUGN-UHFFFAOYSA-N
Synonym phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane
PubChem CID 69849
IUPAC Name trimethyl(phenyl)silane
SMILES C[Si](C)(C)C1=CC=CC=C1

Specifications

Boiling Point 169°C to 170°C
Quantity 25 g
Flash Point 40°C (104°F)
Refractive Index 1.49
Density 0.871
UN Number UN1993
Beilstein 1855012
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 150.3
Percent Purity 98%
Chemical Name or Material Phenyltrimethylsilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-192-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only