missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium tribromide, 97%

Catalog No. p-184266
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160940250 25 g Plastic bottle
AC160941000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160940250 Supplier Thermo Scientific Chemicals Supplier No. 160940250

CAS: 4207-56-1 | C9H14Br3N | 375.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 4207-56-1
Molecular Formula C9H14Br3N
Molecular Weight (g/mol) 375.92
MDL Number MFCD00011789
InChI Key RHXJPCQWYPXLEH-UHFFFAOYSA-N
Synonym mono n,n,n-trimethylbenzenaminium tribromide
PubChem CID 21525000
IUPAC Name trimethyl(phenyl)azanium;triiodide
SMILES C[N+](C)(C)C1=CC=CC=C1.I[I-]I

Specifications

Melting Point 113.0°C to 117.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula C6H5N(CH3)3Br·Br2
Quantity 25 g
Fieser 01,855; 02,328; 04,386
Solubility Information Solubility in water: insoluble.
Formula Weight 375.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenyltrimethylammonium tribromide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 224-127-8
TSCA TSCA

RUO – Research Use Only