Learn More
Phenyltrimethylammonium tribromide, 97%
CAS: 4207-56-1 | C9H14I3N | 516.931 g/mol
Supplier: Thermo Scientific Chemicals A153260B
Description
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.
Solubility
Insoluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.
Specifications
| Phenyltrimethylammonium tribromide | |
| 112°C to 117°C | |
| C9H14I3N | |
| 1000 g | |
| 3919459 | |
| mono n,n,n-trimethylbenzenaminium tribromide | |
| RHXJPCQWYPXLEH-UHFFFAOYSA-N | |
| trimethyl(phenyl)azanium;triiodide | |
| 21525000 | |
| 97% |
| 4207-56-1 | |
| Irritating | |
| MFCD00011789 | |
| UN3261 | |
| Hygroscopic | |
| Insoluble in water. | |
| C[N+](C)(C)C1=CC=CC=C1.I[I-]I | |
| 516.931 | |
| 375.93 |
Chemical Identifiers
| 4207-56-1 | |
| 516.931 | |
| RHXJPCQWYPXLEH-UHFFFAOYSA-N | |
| 21525000 | |
| C[N+](C)(C)C1=CC=CC=C1.I[I-]I |
| C9H14I3N | |
| MFCD00011789 | |
| mono n,n,n-trimethylbenzenaminium tribromide | |
| trimethyl(phenyl)azanium;triiodide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 224-127-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only