missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium tribromide, 97%

Catalog No. AAA1532618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1532618 50 g
AAA1532630 250 g
AAA153260B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1532618 Supplier Thermo Scientific Chemicals Supplier No. A1532618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4207-56-1 | C9H14I3N | 516.931 g/mol

Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.

Chemical Identifiers

CAS 4207-56-1
Molecular Formula C9H14I3N
Molecular Weight (g/mol) 516.931
MDL Number MFCD00011789
InChI Key RHXJPCQWYPXLEH-UHFFFAOYSA-N
Synonym mono n,n,n-trimethylbenzenaminium tribromide
PubChem CID 21525000
IUPAC Name trimethyl(phenyl)azanium;triiodide
SMILES C[N+](C)(C)C1=CC=CC=C1.I[I-]I

Specifications

Melting Point 112°C to 117°C
Odor Irritating
Quantity 50 g
UN Number UN3261
Beilstein 3919459
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 375.93
Percent Purity 97%
Chemical Name or Material Phenyltrimethylammonium tribromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 224-127-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only