Learn More
Phenyltrimethylammonium iodide, 99%
CAS: 98-04-4 | C9H14IN | 263.122 g/mol
Supplier: Thermo Scientific Chemicals A1440236
Description
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water and methanol.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
Specifications
| Phenyltrimethylammonium iodide | |
| ∼227°C (sublimation) | |
| MFCD00011791 | |
| UN2811 | |
| Light sensitive | |
| phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide | |
| KKLAORVGAKUOPZ-UHFFFAOYSA-M | |
| trimethyl(phenyl)azanium;iodide | |
| 94135 | |
| 99% |
| 98-04-4 | |
| C9H14IN | |
| 500 g | |
| 3916752 | |
| 14,7318 | |
| Soluble in water and methanol. | |
| C[N+](C)(C)C1=CC=CC=C1.[I-] | |
| 263.122 | |
| 263.12 |
Chemical Identifiers
| 98-04-4 | |
| 263.122 | |
| KKLAORVGAKUOPZ-UHFFFAOYSA-M | |
| 94135 | |
| C[N+](C)(C)C1=CC=CC=C1.[I-] |
| C9H14IN | |
| MFCD00011791 | |
| phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide | |
| trimethyl(phenyl)azanium;iodide |
Safety and Handling
GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-630-3
RTECSNumber : BT2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only