missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 97%

CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol

Supplier:  Thermo Scientific Chemicals A1382722

 View more versions of this product

Catalog No. AAA1382722


Only null left
Explore available promotions

Description

Description

Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
Specifications

Specifications

210°C to 212°C
-25°C
100 g
MFCD00025689
86°C (186°F)
Reacts with water.
CO[Si](OC)(OC)C1=CC=CC=C1
198.29
198.3
1.4735
1.07
Phenyltrimethoxysilane
2996-92-1
C9H14O3Si
2937896
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
trimethoxy(phenyl)silane
18137
97%
Moisture sensitive

Chemical Identifiers

2996-92-1
198.29
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
18137
CO[Si](OC)(OC)C1=CC=CC=C1
C9H14O3Si
MFCD00025689
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
trimethoxy(phenyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P314-P330-P370+P378q-P501c

H226-H302-H373

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 221-066-9

RTECSNumber : VV5252000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only