missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 97%

Catalog No. AAA1382714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1382714 25 g
AAA1382722 100 g
AAA1382736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1382714 Supplier Thermo Scientific Chemicals Supplier No. A1382714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol

Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.

Chemical Identifiers

CAS 2996-92-1
Molecular Formula C9H14O3Si
Molecular Weight (g/mol) 198.29
MDL Number MFCD00025689
InChI Key ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Synonym phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
PubChem CID 18137
IUPAC Name trimethoxy(phenyl)silane
SMILES CO[Si](OC)(OC)C1=CC=CC=C1

Specifications

Boiling Point 210°C to 212°C
Melting Point -25°C
Quantity 25 g
Beilstein 2937896
Flash Point 86°C (186°F)
Solubility Information Reacts with water.
Formula Weight 198.3
Percent Purity 97%
Refractive Index 1.4735
Sensitivity Moisture sensitive
Density 1.07
Chemical Name or Material Phenyltrimethoxysilane
Show More Show Less
Hazard Category H226-H302-H373
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P314-P330-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-066-9
RTECSNumber VV5252000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only