missing translation for 'onlineSavingsMsg'
Learn More

Phenyltin Trichloride 98.0+%, TCI America™

Catalog No. p-7135051
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
P12825G 5 g
1 options

Catalog No. P12825G

Supplier: TCI America P12825G

Only null left

Chemical Identifiers

CAS 1124-19-2
Molecular Formula C6H5Cl3Sn
Molecular Weight (g/mol) 302.166
MDL Number MFCD00000513
InChI Key UBOGEXSQACVGEC-UHFFFAOYSA-K
Synonym phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
PubChem CID 14298
IUPAC Name trichloro(phenyl)stannane
SMILES C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
UN Number 3265
Formula Weight 302.17
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Phenyltin Trichloride
RTECSNumber WH8730000
TSCA No