missing translation for 'onlineSavingsMsg'
Learn More

Phenylthiohydantoin-arginine Hydrochloride 98.0+%, TCI America™

Catalog No. P0731100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0731100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0731100MG Supplier TCI America Supplier No. P0731100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 182153-75-9
Molecular Formula C13H18ClN5OS
Molecular Weight (g/mol) 327.831
MDL Number MFCD00060177
InChI Key IXKPSQGERWCHMF-UHFFFAOYSA-N
Synonym 5-(3-Guanidinopropyl)-3-phenyl-2-thiohydantoin Hydrochloride, PTH-arginine Hydrochloride
PubChem CID 44630443
IUPAC Name 2-[3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propyl]guanidine;hydrochloride
SMILES C1=CC=C(C=C1)N2C(=O)C(NC2=S)CCCN=C(N)N.Cl

Specifications

Melting Point 188°C
Color Yellow
Quantity 100 mg
Formula Weight 327.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-arginine Hydrochloride