missing translation for 'onlineSavingsMsg'
Learn More

Phenylsuccinic Anhydride 98.0+%, TCI America™

Catalog No. P22575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22575G 5 g
P225725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22575G Supplier TCI America Supplier No. P22575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1131-15-3
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00192395
InChI Key HDFKMLFDDYWABF-UHFFFAOYNA-N
Synonym phenylsuccinic anhydride, 3-phenyldihydrofuran-2,5-dione, monophenyl succinic anhydride, 2,5-furandione, dihydro-3-phenyl, acmc-20ajwd, acmc-20mmqc, phenylsuccinicanhydride, 2,5-furandione, dihydro-3-phenyl-, 3r, phenyl succinic anhydride
PubChem CID 136911
IUPAC Name 3-phenyloxolane-2,5-dione
SMILES O=C1CC(C(=O)O1)C1=CC=CC=C1

Specifications

Melting Point 54°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylsuccinic Anhydride