missing translation for 'onlineSavingsMsg'
Learn More

Phenylsuccinic Acid 98.0+%, TCI America™

Catalog No. P015525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P015525G 25 g
P0155500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P015525G Supplier TCI America Supplier No. P015525G

Chemical Identifiers

CAS 635-51-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004256
InChI Key LVFFZQQWIZURIO-UHFFFAOYSA-N
Synonym phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
PubChem CID 95459
IUPAC Name 2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylsuccinic Acid
EINECSNumber (3)-3108
TSCA Yes