missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane, 97+%

Catalog No. p-4510440
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291690050 5 mL Glass bottle
AC291690250 25 mL Glass bottle
AC291691000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC291690050 Supplier Thermo Scientific Chemicals Supplier No. 291690050
Only null left

CAS: 694-53-1 | C6H8Si | 108.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6H8Si
Molecular Weight (g/mol) 108.22
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Color Colorless
Density 0.8700g/mL
Boiling Point 120.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120
Linear Formula C6H5SiH3
Quantity 5 mL
Beilstein 03,1198
Fieser 05,523; 17,284
Specific Gravity 0.87
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common organic solvents
Formula Weight 108.22
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Phenylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
In contact with water release
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
EINECSNumber 211-772-5

RUO – Research Use Only