Learn More
Phenylsilane, 97%
CAS: 694-53-1 | C6 H8 Si | 108.22 g/mol
Supplier: Thermo Scientific Chemicals A1790114
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.
Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
Specifications
| 120°C | |
| 694-53-1 | |
| C6 H8 Si | |
| UN3399 | |
| phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-, | |
| XJWOWXZSFTXJEX-UHFFFAOYSA-N | |
| phenylsilicon | |
| 6327628 | |
| 97% | |
| Moisture sensitive |
| Phenylsilane | |
| 25 g | |
| MFCD00013478 | |
| 7°C (45°F) | |
| Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents. | |
| C1=CC=C(C=C1)[Si] | |
| 108.22 | |
| 108.22 | |
| 1.5105 | |
| 0.88 |
Chemical Identifiers
| 694-53-1 | |
| 108.22 | |
| XJWOWXZSFTXJEX-UHFFFAOYSA-N | |
| 6327628 | |
| C1=CC=C(C=C1)[Si] |
| C6 H8 Si | |
| MFCD00013478 | |
| phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-, | |
| phenylsilicon |
Safety and Handling
GHS H Statement
H225-H261-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
In contact with water releases flammable gas.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P335+P334-P363-P370+P378q-P50
H225-H261-H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber : 211-772-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only