missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylsilane, 97+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Color | Colorless |
| Density | 0.8700g/mL |
| Boiling Point | 120.0°C |
| Flash Point | 7°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5100 to 1.5120 |
| Linear Formula | C6H5SiH3 |
| Quantity | 25 mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Highly flammable liquid and vapor. Harmful if swallowed. Harmful if inhaled. In contact with water release |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture |
| EINECSNumber | 211-772-5 |
RUO – Research Use Only