missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Phenylsilane
Description
Phenylsilane (PhSiH3) may be used as a reducing agent for the partial reduction of phosphine oxide groups in poly(4,4'sa-diphenylphenylphosphine oxide) (PAPO) to phosphine. It can also undergo photopolymerization with methyl methacrylate(MMA) to form poly(MMA) containing SiH groups. PhSiH3 may be used as a silicon source to synthesize crystalline Si nanowires via Au-nanocrystal-seeded supercritical fluid-liquid-solid (SFLS) synthesis.
Specifications
Specifications
| CAS | 694-53-1 |
| Density | 0.877 g/mL at 25°C |
| Boiling Point | 120°C |
| Molecular Formula | C6H8Si |
| Refractive Index | n20/D 1.510 |
| Linear Formula | C6H5SiH3 |
| MDL Number | MFCD00013478 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 108.21 g/mol |
| Biological Activity | Respiratory System |