Learn More
Phenylselenyl chloride, 98%
CAS: 5707-04-0 | C6H5ClSe | 191.53 g/mol
$275.72 - $275.72
Chemical Identifiers
| CAS | 5707-04-0 |
|---|---|
| Molecular Formula | C6H5ClSe |
| Molecular Weight (g/mol) | 191.53 |
| MDL Number | MFCD00000478 |
| InChI Key | WJCXADMLESSGRI-UHFFFAOYSA-N |
| Synonym | phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene |
| PubChem CID | 21928 |
| IUPAC Name | phenyl selenohypochlorite |
| SMILES | Cl[Se]C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC179510100
|
Thermo Scientific Chemicals
179510100 |
10 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
| 5707-04-0 | |
| 191.53 | |
| WJCXADMLESSGRI-UHFFFAOYSA-N | |
| 21928 | |
| Cl[Se]C1=CC=CC=C1 |
| C6H5ClSe | |
| MFCD00000478 | |
| phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene | |
| phenyl selenohypochlorite |
Specifications
| 5707-04-0 | |
| Orange | |
| Authentic | |
| Glass bottle | |
| C6H5SeCl | |
| 10 g | |
| 05,518; 06,459; 07,286; 09,25; 11,33; 13,26; 16,19; 17,26 | |
| WJCXADMLESSGRI-UHFFFAOYSA-N | |
| phenyl selenohypochlorite | |
| 21928 | |
| 98% | |
| Phenylselenyl chloride, 98% |
| 61.0°C to 65.0°C | |
| 120.0°C (20.0 mmHg) | |
| 97.5% min. (Argentometry) | |
| C6H5ClSe | |
| MFCD00000478 | |
| 06, III, 1110 | |
| phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene | |
| Cl[Se]C1=CC=CC=C1 | |
| 191.53 | |
| 191.52 | |
| Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 227-196-2
RUO – Research Use Only