Learn More
Phenylselenyl bromide, 98%
CAS: 34837-55-3 | C6H5BrSe | 235.97 g/mol
$412.07 - $412.07
Chemical Identifiers
| CAS | 34837-55-3 |
|---|---|
| Molecular Formula | C6H5BrSe |
| Molecular Weight (g/mol) | 235.97 |
| MDL Number | MFCD00000047 |
| InChI Key | LCEFEIBEOBPPSJ-UHFFFAOYSA-N |
| Synonym | phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide |
| PubChem CID | 123446 |
| IUPAC Name | phenyl selenohypobromite |
| SMILES | C1=CC=C(C=C1)[Se]Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC223060250
|
Thermo Scientific Chemicals
223060250 |
25 g | Glass bottle |
Each for $412.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34837-55-3 | |
| 235.97 | |
| LCEFEIBEOBPPSJ-UHFFFAOYSA-N | |
| 123446 | |
| C1=CC=C(C=C1)[Se]Br |
| C6H5BrSe | |
| MFCD00000047 | |
| phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide | |
| phenyl selenohypobromite |
Specifications
| 34837-55-3 | |
| Black to Brown | |
| Authentic | |
| Glass bottle | |
| C6H5SeBr | |
| 25 g | |
| 06,459; 07,286; 11,33; 13,26; 01,1047; 11,487; 16,272 | |
| Solubility in water: insoluble | |
| C1=CC=C(C=C1)[Se]Br | |
| 235.97 | |
| 235.97 | |
| Crystals |
| 60°C to 62°C | |
| 107°C to 108°C (15.0 mmHg) | |
| 98% | |
| C6H5BrSe | |
| MFCD00000047 | |
| 06, II, 319 | |
| phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide | |
| LCEFEIBEOBPPSJ-UHFFFAOYSA-N | |
| phenyl selenohypobromite | |
| 123446 | |
| 98% | |
| Phenylselenyl bromide, 98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effec
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Danger
EINECSNumber : 252-238-1
RUO – Research Use Only