missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl Chloride 97.0+%, TCI America™

Catalog No. P10915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10915G 5 g
P109125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10915G Supplier TCI America Supplier No. P10915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5707-04-0
Molecular Formula C6H5ClSe
Molecular Weight (g/mol) 191.53
MDL Number MFCD00000478
InChI Key WJCXADMLESSGRI-UHFFFAOYSA-N
Synonym phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
PubChem CID 21928
IUPAC Name (phenylselanyl)chlorane
SMILES Cl[Se]C1=CC=CC=C1

Specifications

Melting Point 63°C
Boiling Point 95°C
Quantity 5 g
UN Number 2928
Formula Weight 191.52
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylselenenyl Chloride
TSCA No
Recommended Storage Refrigerator