Learn More
Phenylselenenyl chloride, 98%
CAS: 5707-04-0 | C6H5ClSe | 191.53 g/mol
$154.99 - $564.90
Chemical Identifiers
| CAS | 5707-04-0 |
|---|---|
| Molecular Formula | C6H5ClSe |
| Molecular Weight (g/mol) | 191.53 |
| MDL Number | MFCD00000478 |
| InChI Key | WJCXADMLESSGRI-UHFFFAOYSA-N |
| Synonym | phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene |
| PubChem CID | 21928 |
| IUPAC Name | phenyl selenohypochlorite |
| SMILES | Cl[Se]C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1275106
|
Thermo Scientific Chemicals
A1275106 |
5 g |
Each for $154.99
|
|
|||||
|
AAA1275114
|
Thermo Scientific Chemicals
A1275114 |
25 g |
Each for $564.90
|
|
|||||
Description
Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.
Solubility
Soluble in methanol.
Notes
Moisture sensitive. Incompatible with strong bases, strong oxidizing agents and strong alkalis.
Chemical Identifiers
| 5707-04-0 | |
| 191.53 | |
| WJCXADMLESSGRI-UHFFFAOYSA-N | |
| 21928 | |
| Cl[Se]C1=CC=CC=C1 |
| C6H5ClSe | |
| MFCD00000478 | |
| phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene | |
| phenyl selenohypochlorite |
Specifications
| 5707-04-0 | |
| 120°C (20 mmHg) | |
| C6H5ClSe | |
| 5 g | |
| 1237091 | |
| phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene | |
| WJCXADMLESSGRI-UHFFFAOYSA-N | |
| phenyl selenohypochlorite | |
| 21928 | |
| 98% |
| 60°C to 65°C | |
| Stench | |
| MFCD00000478 | |
| UN2928 | |
| Air and moisture sensitive | |
| Soluble in methanol. | |
| Cl[Se]C1=CC=CC=C1 | |
| 191.53 | |
| 191.52 | |
| Phenylselenenyl chloride |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 227-196-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only